Anti-RRM2 Rabbit Polyclonal Antibody

Lieferant: US Biological
132850
USBI132850EA 662 EUR
USBI132850
Anti-RRM2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RRM2 (ribonucleotide reductase M2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR