Anti-Semaphorin 3G Mouse Polyclonal Antibody
Lieferant: US Biological
133100
USBI133100EA 623 EUR
USBI133100
Anti-Semaphorin 3G Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Semaphorin 3G
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human