Anti-SLC12A1 Mouse Monoclonal Antibody [clone: 4H4]

Lieferant: US Biological
133398
USBI133398EA 623 EUR
USBI133398
Anti-SLC12A1 Mouse Monoclonal Antibody [clone: 4H4]
Antikörper
Type: Primary
Antigen: SLC12A1 (solute carrier family 12 (sodium/potassium/chloride transporter), member 1)
Clonality: monoclonal
Clone: 4H4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR