Anti-SPR Rabbit Polyclonal Antibody

Lieferant: US Biological
133797
USBI133797EA 696 EUR
USBI133797
Anti-SPR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPR (sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR