Anti-SPRR2F Mouse Monoclonal Antibody [clone: 5A9]

Lieferant: US Biological
133802
USBI133802EA 655 EUR
USBI133802
Anti-SPRR2F Mouse Monoclonal Antibody [clone: 5A9]
Antikörper
Type: Primary
Antigen: SPRR2F (small proline-rich protein 2F)
Clonality: monoclonal
Clone: 5A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR