Anti-STAT3 Mouse Polyclonal Antibody

Lieferant: US Biological
133933
USBI133933EA 655 EUR
USBI133933
Anti-STAT3 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR