Anti-SUMO2 Mouse Monoclonal Antibody [clone: 2H8]
Lieferant: US Biological
134091
USBI134091EA 623 EUR
USBI134091
Anti-SUMO2 Mouse Monoclonal Antibody [clone: 2H8]
Antikörper
Recommended dilution: Immunochemistry: 1. 5 ?g/ml
Type: Primary
Antigen: SUMO2 (small ubiquitin-like modifier 2)
Clonality: monoclonal
Clone: 2H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human