Anti-SYT4 Mouse Monoclonal Antibody [clone: 5F8]

Lieferant: US Biological
134151
USBI134151EA 655 EUR
USBI134151
Anti-SYT4 Mouse Monoclonal Antibody [clone: 5F8]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: SYT4 (synaptotagmin IV)
Clonality: monoclonal
Clone: 5F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR