Anti-TEC Mouse Monoclonal Antibody [clone: 1C11]

Lieferant: US Biological
134326
USBI134326EA 655 EUR
USBI134326
Anti-TEC Mouse Monoclonal Antibody [clone: 1C11]
Antikörper
Type: Primary
Antigen: TEC (tec protein tyrosine kinase)
Clonality: monoclonal
Clone: 1C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR