Anti-TRAF2 Rabbit Polyclonal Antibody

Lieferant: US Biological
134667
USBI134667EA 696 EUR
USBI134667
Anti-TRAF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR