Anti-Tastin Mouse Polyclonal Antibody

Lieferant: US Biological
134784
USBI134784EA 655 EUR
USBI134784
Anti-Tastin Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Tastin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR