Anti-Tubulin beta-3 Chain Rabbit Polyclonal Antibody

Lieferant: US Biological
134890
USBI134890EA 662 EUR
USBI134890
Anti-Tubulin beta-3 Chain Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Tubulin beta-3 Chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR