Anti-UEVLD Mouse Polyclonal Antibody

Lieferant: US Biological
135059
USBI135059EA 655 EUR
USBI135059
Anti-UEVLD Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UEVLD (UEV and lactate/malate dehyrogenase domains)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR