Anti-UHRF1BP1L Mouse Polyclonal Antibody

Lieferant: US Biological
135083
USBI135083EA 655 EUR
USBI135083
Anti-UHRF1BP1L Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UHRF1BP1L (UHRF1 binding protein 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR