Anti-UMPS Mouse Polyclonal Antibody

Lieferant: US Biological
135097
USBI135097EA 623 EUR
USBI135097
Anti-UMPS Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UMPS (uridine monophosphate synthetase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR