Anti-WHSC1L1 Mouse Polyclonal Antibody

Lieferant: US Biological
135380
USBI135380EA 623 EUR
USBI135380
Anti-WHSC1L1 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: WHSC1L1 (Wolf-Hirschhorn syndrome candidate 1-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR