Anti-YWHAZ Mouse Monoclonal Antibody [clone: 1B3]

Lieferant: US Biological
135490
USBI135490EA 655 EUR
USBI135490
Anti-YWHAZ Mouse Monoclonal Antibody [clone: 1B3]
Antikörper
Type: Primary
Antigen: YWHAZ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta)
Clonality: monoclonal
Clone: 1B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR