Anti-ZNF398 Mouse Monoclonal Antibody [clone: 4G3]

Lieferant: US Biological
135724
USBI135724EA 655 EUR
USBI135724
Anti-ZNF398 Mouse Monoclonal Antibody [clone: 4G3]
Antikörper
Type: Primary
Antigen: ZNF398 (zinc finger protein 398)
Clonality: monoclonal
Clone: 4G3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR