Anti-HLA-DR Rat Monoclonal Antibody

Lieferant: US Biological
136165
USBI136165EA 632 EUR
USBI136165
Anti-HLA-DR Rat Monoclonal Antibody
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR