Anti-PIAS3 Rabbit Polyclonal Antibody

Lieferant: US Biological
136330
USBI136330EA 632 EUR
USBI136330
Anti-PIAS3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: PIAS3 (protein inhibitor of activated STAT, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR