Anti-Prostaglandin E2 Receptor EP3 Subtype Rabbit Polyclonal Antibody

Lieferant: US Biological
136353
USBI136353EA 675 EUR
USBI136353
Anti-Prostaglandin E2 Receptor EP3 Subtype Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 1-5?g/ml

Type: Primary
Antigen: Prostaglandin E2 Receptor EP3 Subtype
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR