Anti-UBA2 Rabbit Polyclonal Antibody

Lieferant: US Biological
136401
USBI136401EA 681 EUR
USBI136401
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:5000; Western Blot: 1:500-1:1000; Immunochemistry:10 ?g/ml

Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR