Anti-ZMYM2 Rabbit Polyclonal Antibody

Lieferant: US Biological
136536
USBI136536EA 632 EUR
USBI136536
Anti-ZMYM2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ZMYM2 (zinc finger, MYM-type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR