Anti-Aflatoxin B1/2 Mouse Monoclonal Antibody [clone: 13F1]

Lieferant: US Biological
136543
USBI136543EA 714 EUR
USBI136543
Anti-Aflatoxin B1/2 Mouse Monoclonal Antibody [clone: 13F1]
Antikörper
Type: Primary
Antigen: Aflatoxin B1/2 (aflatoxin B1/2)
Clonality: monoclonal
Clone: 13F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR