Anti-CA15-3 Cancer Antigen Mouse Monoclonal Antibody [clone: 13F7]

Lieferant: US Biological
136549
USBI136549EA 878 EUR
USBI136549
Anti-CA15-3 Cancer Antigen Mouse Monoclonal Antibody [clone: 13F7]
Antikörper
Recommended dilution: ELISA:; Western Blot: 1:10000

Type: Primary
Antigen: CA15-3 Cancer Antigen
Clonality: monoclonal
Clone: 13F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR