Anti-CA15-3 Cancer Antigen Mouse Monoclonal Antibody [clone: 13F11]

Lieferant: US Biological
136553
USBI136553EA 714 EUR
USBI136553
Anti-CA15-3 Cancer Antigen Mouse Monoclonal Antibody [clone: 13F11]
Antikörper
Type: Primary
Antigen: CA15-3 Cancer Antigen
Clonality: monoclonal
Clone: 13F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR