Anti-Cancer Antigen CA19-9 Mouse Monoclonal Antibody [clone: 13F13]

Lieferant: US Biological
136555
USBI136555EA 472 EUR
USBI136555
Anti-Cancer Antigen CA19-9 Mouse Monoclonal Antibody [clone: 13F13]
Antikörper
Type: Primary
Antigen: Cancer Antigen CA19-9
Clonality: monoclonal
Clone: 13F13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR