Anti-VAMP 1,2 Mouse Monoclonal Antibody [clone: SP-11]

Lieferant: US Biological
136854
USBI136854EA 465 EUR
USBI136854
Anti-VAMP 1,2 Mouse Monoclonal Antibody [clone: SP-11]
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: parrafin

Type: Primary
Antigen: VAMP 1,2
Clonality: monoclonal
Clone: SP-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR