Anti-HIF1AN Rabbit Polyclonal Antibody

Lieferant: US Biological
136908
USBI136908EA 671 EUR
USBI136908
Anti-HIF1AN Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:100; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR