Anti-Malondialdehyde Goat Polyclonal Antibody

Lieferant: US Biological
136923
USBI136923EA 561 EUR
USBI136923
Anti-Malondialdehyde Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:3000

Type: Primary
Antigen: Malondialdehyde
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR