Anti-Serotonin Transporter Mouse Monoclonal Antibody [clone: 17-7A4]

Lieferant: US Biological
136958
USBI136958EA 537 EUR
USBI136958
Anti-Serotonin Transporter Mouse Monoclonal Antibody [clone: 17-7A4]
Antikörper
Recommended dilution: Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Serotonin Transporter
Clonality: monoclonal
Clone: 17-7A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR