Anti-Interleukin-4, Neutralizing Rabbit Monoclonal Antibody [clone: 13G43]

Lieferant: US Biological
137459
USBI137459EA 586 EUR
USBI137459
Anti-Interleukin-4, Neutralizing Rabbit Monoclonal Antibody [clone: 13G43]
Antikörper
Type: Primary
Antigen: Interleukin-4, Neutralizing
Clonality: monoclonal
Clone: 13G43
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR