Anti-NFkB p65, acetaldehyde Rabbit Monoclonal Antibody [clone: 13G54]

Lieferant: US Biological
137473
USBI137473EA 674 EUR
USBI137473
Anti-NFkB p65, acetaldehyde Rabbit Monoclonal Antibody [clone: 13G54]
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:100

Type: Primary
Antigen: NFkB p65, acetaldehyde
Clonality: monoclonal
Clone: 13G54
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR