Anti-ABCG2 Rabbit Polyclonal Antibody

Lieferant: US Biological
137513
USBI137513EA 552 EUR
USBI137513
Anti-ABCG2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 2.5 ?g/ml; Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: ABCG2 (ATP-binding cassette, sub-family G (WHITE), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR