Anti-C2 Rabbit Polyclonal Antibody

Lieferant: US Biological
137568
USBI137568EA 574 EUR
USBI137568
Anti-C2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1. 25 ?g/ml

Type: Primary
Antigen: C2 (complement component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR