Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: 13H32]

Lieferant: US Biological
137617
USBI137617EA 552 EUR
USBI137617
Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: 13H32]
Antikörper
Do not freeze!

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: monoclonal
Clone: 13H32
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR