Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin))/Texas Red®) [clone: 13H63]

Lieferant: US Biological
137649
USBI137649EA 608 EUR
USBI137649
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin))/Texas Red®) [clone: 13H63]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: 13H63
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR