Anti-IL2RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 13H94]

Lieferant: US Biological
137680
USBI137680EA 678 EUR
USBI137680
Anti-IL2RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 13H94]
Antikörper
Do not freeze!

Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: 13H94
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR