Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin))/Cy5.5®) [clone: 13H119]

Lieferant: US Biological
137706
USBI137706EA 630 EUR
USBI137706
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin))/Cy5.5®) [clone: 13H119]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone: 13H119
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR