Anti-CD4a Mouse Monoclonal Antibody (PE (Phycoerythrin))/Cy5®) [clone: 13H161]

Lieferant: US Biological
137752
USBI137752EA 654 EUR
USBI137752
Anti-CD4a Mouse Monoclonal Antibody (PE (Phycoerythrin))/Cy5®) [clone: 13H161]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD4a
Clonality: monoclonal
Clone: 13H161
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR