Anti-CFI Goat Polyclonal Antibody

Lieferant: US Biological
137893
USBI137893EA 473 EUR
USBI137893
Anti-CFI Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:4000-1:8000; Western Blot: 1:2000-1:4000; ImmunodImmunofluorescencefusion: 1:8

Type: Primary
Antigen: CFI (complement factor I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR