Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 13H250]

Lieferant: US Biological
137964
USBI137964EA 977 EUR
USBI137964
Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 13H250]
Antikörper
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 13H250
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR