Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 13H251]

Lieferant: US Biological
137965
USBI137965EA 1020 EUR
USBI137965
Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 13H251]
Antikörper
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 13H251
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR