Anti-HBZ Rabbit Polyclonal Antibody

Lieferant: US Biological
138010
USBI138010EA 784 EUR
USBI138010
Anti-HBZ Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1. 25 ?g/ml; Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: HBZ (hemoglobin, zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR