Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: 13H267]

Lieferant: US Biological
138018
USBI138018EA 452 EUR
USBI138018
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: 13H267]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 13H267
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR