Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: 13H268]

Lieferant: US Biological
138019
USBI138019EA 708 EUR
USBI138019
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: 13H268]
Antikörper
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 13H268
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR