Anti-Protein A Goat Polyclonal Antibody Rhodamine
Lieferant: US Biological
138260
USBI138260EA 479 EUR
USBI138260
Anti-Protein A Goat Polyclonal Antibody Rhodamine
Antikörper
Recommended dilution: Immunofluorescence: 1:500-1:2500
Type: Tag
Antigen: Protein A
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen