Anti-SLC25A16 Rabbit Polyclonal Antibody
Lieferant: US Biological
138358
USBI138358EA 785 EUR
USBI138358
Anti-SLC25A16 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 5 ?g/ml
Type: Primary
Antigen: SLC25A16 (solute carrier family 25 (mitochondrial carrier; Graves disease autoantigen), member 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat