Anti-EPCAM Mouse Monoclonal Antibody [clone: GZ-20]
Lieferant: US Biological
138542-500
USBI138542-500EA 391 EUR
USBI138542-500
Anti-EPCAM Mouse Monoclonal Antibody [clone: GZ-20]
Antikörper
Recommended dilution: Western Blot: 1:100-1:200; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:50-1:100; Immunoprecipitation: 5-10?l/500 ?g
Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: monoclonal
Clone: GZ-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat