Anti-DNASE1 Mouse Monoclonal Antibody [clone: 13j29]

Lieferant: US Biological
140050
USBI140050EA 527 EUR
USBI140050
Anti-DNASE1 Mouse Monoclonal Antibody [clone: 13j29]
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: DNASE1 (deoxyribonuclease I)
Clonality: monoclonal
Clone: 13j29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR